Learn More
Hampton Research SLICE PH REAGENT #17 (B5)
SDP
Catalog No. 50562875
Encompass
HR2-070-17/Slice pH #17 (B5), 185 ml - 1.0 M DL-Malic acid pH 5.6. Please note items from this supplier are non-returnable.
Catalog No. 50-562-875 Supplier Hampton Research Supplier No. HR207017
HR2-070-17/Slice pH #17 (B5), 185 ml - 1.0 M DL-Malic acid pH 5.6. Please note items from this supplier are non-returnable.