Learn More
Hampton Research SLICE PH REAGENT #23 (B11)
SDP
Catalog No. 50562881
Encompass
HR2-070-23/Slice pH #23 (B11), 185 ml - 1.0 M Succinic acid pH 6.0. Please note items from this supplier are non-returnable.
Catalog No. 50-562-881 Supplier Hampton Research Supplier No. HR207023
HR2-070-23/Slice pH #23 (B11), 185 ml - 1.0 M Succinic acid pH 6.0. Please note items from this supplier are non-returnable.