Learn More
Hampton Research SLICE PH REAGENT #25 (C1)
SDP
Supplier:  Hampton Research HR207025
Encompass
HR2-070-25/Slice pH #25 (C1), 185 ml - 1.0 M Sodium cacodylate trihydrate pH 5.5. Please note items from this supplier are non-returnable.
Catalog No. 50-562-883