Learn More
Hampton Research SLICE PH REAGENT #28 (C4)
SDP
Catalog No. 50562886
Encompass
HR2-070-28/Slice pH #28 (C4), 185 ml - 1.0 M Sodium cacodylate trihydrate pH 6.4. Please note items from this supplier are non-returnable.
Catalog No. 50-562-886 Supplier Hampton Research Supplier No. HR207028
HR2-070-28/Slice pH #28 (C4), 185 ml - 1.0 M Sodium cacodylate trihydrate pH 6.4. Please note items from this supplier are non-returnable.