Learn More
Hampton Research SLICE PH REAGENT #29 (C5)
SDP
Catalog No. 50562887
Encompass
HR2-070-29/Slice pH #29 (C5), 185 ml - 1.0 M MES monohydrate pH 5.3. Please note items from this supplier are non-returnable.
Catalog No. 50-562-887 Supplier Hampton Research Supplier No. HR207029
HR2-070-29/Slice pH #29 (C5), 185 ml - 1.0 M MES monohydrate pH 5.3. Please note items from this supplier are non-returnable.