Learn More
Hampton Research SLICE PH REAGENT #30 (C6)
SDP
Catalog No. 50562888
Encompass
HR2-070-30/Slice pH #30 (C6), 185 ml - 1.0 M MES monohydrate pH 5.6. Please note items from this supplier are non-returnable.
Catalog No. 50-562-888 Supplier Hampton Research Supplier No. HR207030
HR2-070-30/Slice pH #30 (C6), 185 ml - 1.0 M MES monohydrate pH 5.6. Please note items from this supplier are non-returnable.