Learn More
Hampton Research SLICE PH REAGENT #32 (C8)
SDP
Catalog No. 50562890
Encompass
HR2-070-32/Slice pH #32 (C8), 185 ml - 1.0 M MES monohydrate pH 6.2. Please note items from this supplier are non-returnable.
Catalog No. 50-562-890 Supplier Hampton Research Supplier No. HR207032
HR2-070-32/Slice pH #32 (C8), 185 ml - 1.0 M MES monohydrate pH 6.2. Please note items from this supplier are non-returnable.