Learn More
Hampton Research SLICE PH REAGENT #33 (C9)
SDP
Catalog No. 50562891
Encompass
HR2-070-33/Slice pH #33 (C9), 185 ml - 1.0 M MES monohydrate pH 6.5. Please note items from this supplier are non-returnable.
Catalog No. 50-562-891 Supplier Hampton Research Supplier No. HR207033
HR2-070-33/Slice pH #33 (C9), 185 ml - 1.0 M MES monohydrate pH 6.5. Please note items from this supplier are non-returnable.