Learn More
Hampton Research SLICE PH REAGENT #34 (C10)
SDP
Catalog No. 50562892
Encompass
HR2-070-34/Slice pH #34 (C10), 185 ml - 1.0 M BIS-TRIS pH 5.7. Please note items from this supplier are non-returnable.
Catalog No. 50-562-892 Supplier Hampton Research Supplier No. HR207034
HR2-070-34/Slice pH #34 (C10), 185 ml - 1.0 M BIS-TRIS pH 5.7. Please note items from this supplier are non-returnable.