Learn More
Hampton Research SLICE PH REAGENT #37 (D1)
SDP
Catalog No. 50562895
Encompass
HR2-070-37/Slice pH #37 (D1), 185 ml - 1.0 M BIS-TRIS pH 6.6. Please note items from this supplier are non-returnable.
Catalog No. 50-562-895 Supplier Hampton Research Supplier No. HR207037
HR2-070-37/Slice pH #37 (D1), 185 ml - 1.0 M BIS-TRIS pH 6.6. Please note items from this supplier are non-returnable.