Learn More
Hampton Research SLICE PH REAGENT #47 (D11)
SDP
Catalog No. 50562905
Encompass
HR2-070-47/Slice pH #47 (D11), 185 ml - 1.0 M Imidazole pH 7.1. Please note items from this supplier are non-returnable.
Catalog No. 50-562-905 Supplier Hampton Research Supplier No. HR207047
HR2-070-47/Slice pH #47 (D11), 185 ml - 1.0 M Imidazole pH 7.1. Please note items from this supplier are non-returnable.