Learn More
Hampton Research SLICE PH REAGENT #48 (D12)
SDP
Catalog No. 50562906
Encompass
HR2-070-48/Slice pH #48 (D12), 185 ml - 1.0 M Imidazole pH 7.4. Please note items from this supplier are non-returnable.
Catalog No. 50-562-906 Supplier Hampton Research Supplier No. HR207048
HR2-070-48/Slice pH #48 (D12), 185 ml - 1.0 M Imidazole pH 7.4. Please note items from this supplier are non-returnable.