Learn More
Hampton Research SLICE PH REAGENT #49 (E1)
SDP
Catalog No. 50562907
Encompass
HR2-070-49/Slice pH #49 (E1), 185 ml - 1.0 M BIS-TRIS propane pH 6.4. Please note items from this supplier are non-returnable.
Catalog No. 50-562-907 Supplier Hampton Research Supplier No. HR207049
HR2-070-49/Slice pH #49 (E1), 185 ml - 1.0 M BIS-TRIS propane pH 6.4. Please note items from this supplier are non-returnable.