Learn More
Hampton Research SLICE PH REAGENT #56 (E8)
SDP
Catalog No. 50562914
Encompass
HR2-070-56/Slice pH #56 (E8), 185 ml - 1.0 M MOPS pH 7.4. Please note items from this supplier are non-returnable.
Catalog No. 50-562-914 Supplier Hampton Research Supplier No. HR207056
HR2-070-56/Slice pH #56 (E8), 185 ml - 1.0 M MOPS pH 7.4. Please note items from this supplier are non-returnable.