Learn More
Hampton Research SLICE PH REAGENT #57 (E9)
SDP
Catalog No. 50562915
Encompass
HR2-070-57/Slice pH #57 (E9), 185 ml - 1.0 M MOPS pH 7.7. Please note items from this supplier are non-returnable.
Catalog No. 50-562-915 Supplier Hampton Research Supplier No. HR207057
HR2-070-57/Slice pH #57 (E9), 185 ml - 1.0 M MOPS pH 7.7. Please note items from this supplier are non-returnable.