Learn More
Hampton Research SLICE PH REAGENT #64 (F4)
SDP
Catalog No. 50562922
Encompass
HR2-070-64/Slice pH #64 (F4), 185 ml - 1.0 M HEPES pH 7.4. Please note items from this supplier are non-returnable.
Catalog No. 50-562-922 Supplier Hampton Research Supplier No. HR207064
HR2-070-64/Slice pH #64 (F4), 185 ml - 1.0 M HEPES pH 7.4. Please note items from this supplier are non-returnable.