Learn More
Hampton Research SLICE PH REAGENT #68 (F8)
SDP
Catalog No. 50562926
Encompass
HR2-070-68/Slice pH #68 (F8), 185 ml - 1.0 M TRIS hydrochloride pH 7.8. Please note items from this supplier are non-returnable.
Catalog No. 50-562-926 Supplier Hampton Research Supplier No. HR207068
HR2-070-68/Slice pH #68 (F8), 185 ml - 1.0 M TRIS hydrochloride pH 7.8. Please note items from this supplier are non-returnable.