Learn More
Hampton Research SLICE PH REAGENT #72 (F12)
SDP
Catalog No. 50562930
Encompass
HR2-070-72/Slice pH #72 (F12), 185 ml - 1.0 M Tris pH 7.9. Please note items from this supplier are non-returnable.
Catalog No. 50-562-930 Supplier Hampton Research Supplier No. HR207072
HR2-070-72/Slice pH #72 (F12), 185 ml - 1.0 M Tris pH 7.9. Please note items from this supplier are non-returnable.