Learn More
Hampton Research SLICE PH REAGENT #73 (G1)
SDP
Catalog No. 50562931
Encompass
HR2-070-73/Slice pH #73 (G1), 185 ml - 1.0 M Tris pH 8.2. Please note items from this supplier are non-returnable.
Catalog No. 50-562-931 Supplier Hampton Research Supplier No. HR207073
HR2-070-73/Slice pH #73 (G1), 185 ml - 1.0 M Tris pH 8.2. Please note items from this supplier are non-returnable.