Learn More
Hampton Research SLICE PH REAGENT #76 (G4)
SDP
Catalog No. 50562934
Encompass
HR2-070-76/Slice pH #76 (G4), 185 ml - 1.0 M Tricine pH 7.7. Please note items from this supplier are non-returnable.
Catalog No. 50-562-934 Supplier Hampton Research Supplier No. HR207076
HR2-070-76/Slice pH #76 (G4), 185 ml - 1.0 M Tricine pH 7.7. Please note items from this supplier are non-returnable.