Learn More
Hampton Research SLICE PH REAGENT #80 (G8)
SDP
Catalog No. 50562938
Encompass
HR2-070-80/Slice pH #80 (G8), 185 ml - 1.0 M BICINE pH 7.5. Please note items from this supplier are non-returnable.
Catalog No. 50-562-938 Supplier Hampton Research Supplier No. HR207080
HR2-070-80/Slice pH #80 (G8), 185 ml - 1.0 M BICINE pH 7.5. Please note items from this supplier are non-returnable.