Learn More
Hampton Research SLICE PH REAGENT #81 (G9)
SDP
Catalog No. 50562939
Encompass
HR2-070-81/Slice pH #81 (G9), 185 ml - 1.0 M BICINE pH 7.8. Please note items from this supplier are non-returnable.
Catalog No. 50-562-939 Supplier Hampton Research Supplier No. HR207081
HR2-070-81/Slice pH #81 (G9), 185 ml - 1.0 M BICINE pH 7.8. Please note items from this supplier are non-returnable.