Learn More
Hampton Research SLICE PH REAGENT #84 (G12)
SDP
Catalog No. 50562942
Encompass
HR2-070-84/Slice pH #84 (G12), 185 ml - 1.0 M BICINE pH 8.7. Please note items from this supplier are non-returnable.
Catalog No. 50-562-942 Supplier Hampton Research Supplier No. HR207084
HR2-070-84/Slice pH #84 (G12), 185 ml - 1.0 M BICINE pH 8.7. Please note items from this supplier are non-returnable.