Learn More
Hampton Research SLICE PH REAGENT #88 (H4)
SDP
Catalog No. 50562946
Encompass
HR2-070-88/Slice pH #88 (H4), 185 ml - 1.0 M BIS-TRIS propane pH 9.4. Please note items from this supplier are non-returnable.
Catalog No. 50-562-946 Supplier Hampton Research Supplier No. HR207088
HR2-070-88/Slice pH #88 (H4), 185 ml - 1.0 M BIS-TRIS propane pH 9.4. Please note items from this supplier are non-returnable.