Learn More
Hampton Research SLICE PH REAGENT #91 (H7)
SDP
Catalog No. 50562949
Encompass
HR2-070-91/Slice pH #91 (H7), 185 ml - 1.0 M Glycine pH 9.2. Please note items from this supplier are non-returnable.
Catalog No. 50-562-949 Supplier Hampton Research Supplier No. HR207091
HR2-070-91/Slice pH #91 (H7), 185 ml - 1.0 M Glycine pH 9.2. Please note items from this supplier are non-returnable.