Learn More
Hampton Research SLICE PH REAGENT #92 (H8)
SDP
Catalog No. 50562950
Encompass
HR2-070-92/Slice pH #92 (H8), 185 ml - 1.0 M Glycine pH 9.5. Please note items from this supplier are non-returnable.
Catalog No. 50-562-950 Supplier Hampton Research Supplier No. HR207092
HR2-070-92/Slice pH #92 (H8), 185 ml - 1.0 M Glycine pH 9.5. Please note items from this supplier are non-returnable.