Learn More
Electron Microscopy Sciences Globe Slides Two-Sided Frosted, Beveled Edge, Clipped Corner
SDP
Catalog No. 501928796
Encompass_Preferred
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Beveled
Corner: Clipped
Catalog No. 50-192-8796 Supplier Electron Microscopy Sciences Supplier No. 7188329
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Beveled
Corner: Clipped