Learn More
Ibidi USA µ-Slide 18 Well, Uncoated: #1.5 polymer coverslip, hydrophobic, sterilized
SDP
Catalog No. NC1826745
Encompass
All-in-one 18 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. A microscopy slide that is ideal for high-resolution imaging through the No. 1.5 polymer coverslip bottom with the highest optical quality. A cell culture chamber suitable for various microscopy techniques.
This item is not returnable. View return policy
Catalog No. NC1826745 Supplier Ibidi USA Supplier No. 81811
All-in-one 18 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. A microscopy slide that is ideal for high-resolution imaging through the No. 1.5 polymer coverslip bottom with the highest optical quality. A cell culture chamber suitable for various microscopy techniques.