Learn More
Ace Glass, Inc. 25mm, body only, slide adapter that enables depth adjustment in all of our reaction vessels.
SDP
Catalog No. 50977145
Encompass_Preferred
25mm, body only, slide adapter that enables depth adjustment in all of our reaction vessels.
Catalog No. 50-977-145 Supplier Ace Glass, Inc. Supplier No. 985221
25mm, body only, slide adapter that enables depth adjustment in all of our reaction vessels.