Learn More
Ace Glass, Inc. Slide adapter, 25mm, complete with bushing
SDP
Catalog No. 50977147
Encompass_Preferred
25mm slide adapter that enables depth adjustment in all of our reaction vessels. Accepts 1/2in horns in #25 Ace-Threds™
Catalog No. 50-977-147 Supplier Ace Glass, Inc. Supplier No. 985241
25mm slide adapter that enables depth adjustment in all of our reaction vessels. Accepts 1/2in horns in #25 Ace-Threds™