Learn More
Ace Glass, Inc. 35mm, body only, slide adapter that enables depth adjustment in all of our reaction vessels.
SDP
Catalog No. 50977146
Encompass_Preferred
35mm, body only, slide adapter that enables depth adjustment in all of our reaction vessels.
Catalog No. 50-977-146 Supplier Ace Glass, Inc. Supplier No. 985225
35mm, body only, slide adapter that enables depth adjustment in all of our reaction vessels.