Learn More
Ace Glass, Inc. Slide adapter, 35mm, complete with bushing
SDP
Catalog No. 50980564
Encompass_Preferred
35mm slide adapter that enables depth adjustment in all of our reaction vessels. Accepts our 3/4 & 1 inch horns in #36 Ace-Threds™
Catalog No. 50-980-564 Supplier Ace Glass, Inc. Supplier No. 985245
35mm slide adapter that enables depth adjustment in all of our reaction vessels. Accepts our 3/4 & 1 inch horns in #36 Ace-Threds™