Learn More
Electron Microscopy Sciences Diamond Etch™ Adhesion Slide, Blue
SDP
Supplier:  Electron Microscopy Sciences 7186502
Encompass_Preferred
Diamond Etch slides, 90 degrees offer optimized etched labeling resolution. Made of white glass designed with top/bottom contrasting colors, specifically for slide etching instruments. Exhibit higher visibility with contrasting background.
Color: Blue
Catalog No. 50-192-8572