Learn More
Electron Microscopy Sciences Millennia™ Adhesion Slide, Burnt Orange, 75 x 25 mm
SDP
Catalog No. 501928565
Encompass_Preferred
Millennia 2.0 Slides have a higher density of binding site, which significantly increases the adhesive forces between the slide and the anchored tissue section. Ideal for immunohistochemistry and can tolerate harsh chemicals with no undesirable background staining, tissue artifact, or specimen loss.
Color: Burnt Orange
Size: 75 x 25 mm
Catalog No. 50-192-8565 Supplier Electron Microscopy Sciences Supplier No. 7186304
Millennia 2.0 Slides have a higher density of binding site, which significantly increases the adhesive forces between the slide and the anchored tissue section. Ideal for immunohistochemistry and can tolerate harsh chemicals with no undesirable background staining, tissue artifact, or specimen loss.
Color: Burnt Orange
Size: 75 x 25 mm