Learn More
Electron Microscopy Sciences Millennia™ Adhesion Slide, Tan, 75 x 25 mm
SDP
Catalog No. 501928569
Encompass_Preferred
Millennia 2.0 Slides have a higher density of binding site, which significantly increases the adhesive forces between the slide and the anchored tissue section. Ideal for immunohistochemistry and can tolerate harsh chemicals with no undesirable background staining, tissue artifact, or specimen loss.
Color: Tan
Size: 75 x 25 mm
Catalog No. 50-192-8569 Supplier Electron Microscopy Sciences Supplier No. 7186308
Millennia 2.0 Slides have a higher density of binding site, which significantly increases the adhesive forces between the slide and the anchored tissue section. Ideal for immunohistochemistry and can tolerate harsh chemicals with no undesirable background staining, tissue artifact, or specimen loss.
Color: Tan
Size: 75 x 25 mm