Learn More
Electron Microscopy Sciences Millennia™ Adhesion Slide, Yellow, 75 x 25 mm
SDP
Catalog No. 501928570
Encompass_Preferred
Millennia 2.0 Slides have a higher density of binding site, which significantly increases the adhesive forces between the slide and the anchored tissue section. Ideal for immunohistochemistry and can tolerate harsh chemicals with no undesirable background staining, tissue artifact, or specimen loss.
Color: Yellow
Size: 75 x 25 mm
Catalog No. 50-192-8570 Supplier Electron Microscopy Sciences Supplier No. 7186309
Millennia 2.0 Slides have a higher density of binding site, which significantly increases the adhesive forces between the slide and the anchored tissue section. Ideal for immunohistochemistry and can tolerate harsh chemicals with no undesirable background staining, tissue artifact, or specimen loss.
Color: Yellow
Size: 75 x 25 mm