Learn More
Andwin Scientific SLIDE BASKET ADAPTER
SDP
Catalog No. NC0213316
Encompass_Preferred
The Prisma 20-Slide Basket adapter is an accessory for Tissue-Tek Prisma, Film and G2 Coverslippers, a ready-to-use product that offers superior performance, convenience, and flexibility in staining and glass coverslipping. This product by Sakura features a quick action of coverslipping 360 slides per hour. It is sold separately. The slide basket adapter is sold individually.
Catalog No. NC0213316 Supplier Andwin Scientific Supplier No. 6136
The Prisma 20-Slide Basket adapter is an accessory for Tissue-Tek Prisma, Film and G2 Coverslippers, a ready-to-use product that offers superior performance, convenience, and flexibility in staining and glass coverslipping. This product by Sakura features a quick action of coverslipping 360 slides per hour. It is sold separately. The slide basket adapter is sold individually.