Learn More
Electron Microscopy Sciences Globe Plain Slides, Beveled Edge, Clipped Corner
SDP
Supplier:  Electron Microscopy Sciences 7188318
Encompass_Preferred
These plain microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings. These slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1.1 mm
Edge: Beveled
Corner: Clipped
Catalog No. 50-192-8785