Learn More
Med Tek Corporation Disposo-Type Blood Typing Slide, 3 Deep Wells, Non-Sterile, 2 x 3 in
Catalog No. NC2259419
Encompass_Preferred
The Disposo-Type Blood Typing Slide is designed for efficient ABO and Rh blood typing in laboratories. Made of durable styrene plastic, it features three deep, oval wells for precise sample mixing. Its clear bold markings help minimize testing errors, ensuring reliable agglutination observation. With 432 slides per case, this disposable, non-sterile product streamlines blood typing procedures, making it essential for medical and educational settings.
Catalog No. NC2259419 Supplier Med Tek Corporation Supplier No. 7013 CONCAVX3 BOXES
The Disposo-Type Blood Typing Slide is designed for efficient ABO and Rh blood typing in laboratories. Made of durable styrene plastic, it features three deep, oval wells for precise sample mixing. Its clear bold markings help minimize testing errors, ensuring reliable agglutination observation. With 432 slides per case, this disposable, non-sterile product streamlines blood typing procedures, making it essential for medical and educational settings.