Learn More
Med Tek Corporation Med Tek Blood Typing Slide, Disposable, Oval Wells, 864/CS
Catalog No. NC2228378
Encompass_Preferred
The disposable blood typing slide by Med Tek Corporation features three deep, oval-shaped wells designed for accurate blood mixing without spillage. Constructed from durable styrene plastic, it ensures users can easily identify blood types through clearly marked bold letters, minimizing errors during laboratory procedures. This product stands out for its effective design and reliability in clinical settings, enabling precise blood typing for enhanced laboratory efficiency.
This item is not returnable. View return policy
Catalog No. NC2228378 Supplier Med Tek Corporation Supplier No. 7013 CONCAVXCS
The disposable blood typing slide by Med Tek Corporation features three deep, oval-shaped wells designed for accurate blood mixing without spillage. Constructed from durable styrene plastic, it ensures users can easily identify blood types through clearly marked bold letters, minimizing errors during laboratory procedures. This product stands out for its effective design and reliability in clinical settings, enabling precise blood typing for enhanced laboratory efficiency.