Learn More
Med Vet International Bel-Art Products 44577-0000, Slide Box For 100 Slides With Hinged Lid, Each
Catalog No. 502829672
Encompass_Preferred
Slide Box for 100 Slides with Hinged Lid Supreme defense for standard 25 x 75mm (1" x 3") microscope slides. Features: Heavy-walled, moisture- and insect-proof Slim, durable design Easily identify slides with included numbered index on base and cover Box walls are rugged and won't bend, split, or chip Storage box for holding microscope slides securely Holds 25 x 75mm (1 x 3") microscope slides (sold separately)Numbered slide positions and content sheet for listing slides Polystyrene box construction for durability
Catalog No. 50-282-9672 Supplier Med Vet International Supplier No. 445770000
Slide Box for 100 Slides with Hinged Lid Supreme defense for standard 25 x 75mm (1" x 3") microscope slides. Features: Heavy-walled, moisture- and insect-proof Slim, durable design Easily identify slides with included numbered index on base and cover Box walls are rugged and won't bend, split, or chip Storage box for holding microscope slides securely Holds 25 x 75mm (1 x 3") microscope slides (sold separately)Numbered slide positions and content sheet for listing slides Polystyrene box construction for durability