Learn More
Research Products International Corp Microscope Slide Box, 100 Slide Capacity, White, Foam Lining, 208 x 175 x 34 mm
SDP
Catalog No. 501367716
Encompass
Slide boxes are perfect for storage or transportation of standard size slides (25 x 75 mm and 1 x 3 inch). Molded, rigid boxes securely hold slides in grooved slots. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Closes securely with a rust-resistant, nickel-plated clasp and pin hinges.
Catalog No. 50-136-7716 Supplier Research Products International Corp Supplier No. 247720
Slide boxes are perfect for storage or transportation of standard size slides (25 x 75 mm and 1 x 3 inch). Molded, rigid boxes securely hold slides in grooved slots. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Closes securely with a rust-resistant, nickel-plated clasp and pin hinges.