Learn More
Thomas Scientific 100-Place Slide Box, Blue, Foam
SDP
Catalog No. NC9013915
Encompass_Preferred
Slide boxes are perfect for storage or transportation of standard size slides (25 x 75 mm and 1 x 3"). Molded, rigid boxes securely hold slides in grooved slots Durable ABS plastic boxes are available with either cork or foam lining Both the lining and the slots are numbered to correspond to the slide inventory sheet Close securely with a rust-resistant, nickel-plated clasp and pin hinges L x W x H: 208 x 175 x 34 mm
Catalog No. NC9013915 Supplier Thomas Scientific Supplier No. 6708F50
Slide boxes are perfect for storage or transportation of standard size slides (25 x 75 mm and 1 x 3"). Molded, rigid boxes securely hold slides in grooved slots Durable ABS plastic boxes are available with either cork or foam lining Both the lining and the slots are numbered to correspond to the slide inventory sheet Close securely with a rust-resistant, nickel-plated clasp and pin hinges L x W x H: 208 x 175 x 34 mm