Learn More
Sigma Aldrich Fine Chemicals Biosciences Slide box 25 place blue
Catalog No. 502900443
Encompass
Standard Microscope Slide Boxes Constructed of durable polypropyleneSlide boxes hold either 25 or 100 standard slides (25 x 75 mm and 1 x 3 inches).Hinged lids are held securely closed with a latch lockStacking feet maximize stabilitySlide identification card - Capacity 100 slides Hinge Latch Lock 210 x 169 x 37 mm Blue
Catalog No. 50-290-0443 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS15990A
Standard Microscope Slide Boxes Constructed of durable polypropyleneSlide boxes hold either 25 or 100 standard slides (25 x 75 mm and 1 x 3 inches).Hinged lids are held securely closed with a latch lockStacking feet maximize stabilitySlide identification card - Capacity 100 slides Hinge Latch Lock 210 x 169 x 37 mm Blue