Learn More
Research Products International Corp Heavy Duty Microscope Slide Box, Holds Fifty 1 x 3 Inch Slides
SDP
Catalog No. 50550280
Encompass
Durable impact resistant plastic. Durable microscope slide box features a snug fitting removable cover and thick walls that resist bending and cracking. Unlined base and snug fitting lid provides a moisture resistant interior. Base has fifty individually numbered slots that accommodate 1" x 3" slides. A numbered index card is also included that mounts inside the cover. Boxes also feature molded in stacking notches and label area.
Catalog No. 50-550-280 Supplier Research Products International Corp Supplier No. 247684
Durable impact resistant plastic. Durable microscope slide box features a snug fitting removable cover and thick walls that resist bending and cracking. Unlined base and snug fitting lid provides a moisture resistant interior. Base has fifty individually numbered slots that accommodate 1" x 3" slides. A numbered index card is also included that mounts inside the cover. Boxes also feature molded in stacking notches and label area.