Learn More
Newcomer Supply Slide Brite Xylene Substitute, 4x1 gallon case. It is Odorless!
SDP
Catalog No. NC0795640
Encompass
An extremely safe, odorless alternative to xylene. Gentle on tissue and does not make it brittle. Excellent for derms and biopsy tissue. No tissue shrinkage or morphology changes. Enhances nuclear detail. Non-flammable with a flash point of 142°F. Use with S-Mount Mounting Medium for optimal results. No hazardous shipping fees!
Catalog No. NC0795640 Supplier Newcomer Supply Supplier No. ABSB04
An extremely safe, odorless alternative to xylene. Gentle on tissue and does not make it brittle. Excellent for derms and biopsy tissue. No tissue shrinkage or morphology changes. Enhances nuclear detail. Non-flammable with a flash point of 142°F. Use with S-Mount Mounting Medium for optimal results. No hazardous shipping fees!