Learn More
StatLab Slide Brite Xylene Substitute
SDP
Catalog No. 21033453
Change view
Click to view available options
Product Type:
Xylene Substitute
Catalog No. Product Type
21-033-453 Xylene Substitute
1 options
Catalog No. 21-033-453 Supplier StatLab Supplier No. SB04
Only null left
Add to Cart
Add to Cart
Aliphatic hydrocarbon for processing and staining
  • A high purity, odorless, fast-drying, non-irritating, xylene substitute
  • Low Odor
  • Flashpoint is 145F
Specifications
Product Type Xylene Substitute