Learn More
Electron Microscopy Sciences Retriever Thermal Slide Processor Chamber
SDP
Catalog No. 5031171
Encompass_Preferred
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.
Catalog No. 50-311-71 Supplier Electron Microscopy Sciences Supplier No. 6270501
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.