Learn More
Electron Microscopy Sciences Globe White Glass Charged Slides, 90° Ground Edge, Corner, Pink Frosted Label
SDP
Catalog No. 501928860
Encompass_Preferred
Produced from the finest quality optical white glass, these charged microscope slides provide the ideal surface for cell and tissue adhesion. They are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. The slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1-1.2 mm
Edge: 90° Ground
Label: Pink Frosted
Catalog No. 50-192-8860 Supplier Electron Microscopy Sciences Supplier No. 7188393
Produced from the finest quality optical white glass, these charged microscope slides provide the ideal surface for cell and tissue adhesion. They are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. The slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1-1.2 mm
Edge: 90° Ground
Label: Pink Frosted