Learn More
Electron Microscopy Sciences Globe White Glass Charged Slides, 90° Ground Edge, Corner, Aqua Frosted Label
SDP
Catalog No. 501928848
Encompass_Preferred
Produced from the finest quality optical white glass, these charged microscope slides provide the ideal surface for cell and tissue adhesion. They are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. The slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1-1.2 mm
Edge: 90° Ground
Label: Aqua Frosted
Catalog No. 50-192-8848 Supplier Electron Microscopy Sciences Supplier No. 7188381
Produced from the finest quality optical white glass, these charged microscope slides provide the ideal surface for cell and tissue adhesion. They are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. The slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1-1.2 mm
Edge: 90° Ground
Label: Aqua Frosted